Learn More
StatLab Mini Elite Fine Point PAP Pen
SDP
Catalog No. 22000004
Change view
Click to view available options
Type:
Permanent Marking Pen
Catalog No. Type
22-000-004 Permanent Marking Pen
1 options
Catalog No. 22-000-004 Supplier StatLab Supplier No. ACR016
Add to Cart
Add to Cart
A fine point liquid blocking PAP pen
  • Suitable for both immunohistochemical and fluorescent staining
  • Hydrophobic properties allow the user to draw barriers on the slide, to confine the flow of reagents to a defined area
Specifications
Type Permanent Marking Pen