Learn More
Research Products International Corp Mini-Environmental Meter
SDP
Supplier:  Research Products International Corp 850070
Encompass
Combines a thermometer, humidity meter, light meter and anemometer in one compact unit. Provides convenient environmental monitoring anywhere. Use to check fume hoods, monitor room temperature and humidity or light output. Supplied complete with carrying case, battery and instructions.
Catalog No. 50-197-7521