Learn More
Mopec™ Mini Formaway Formalin Neutralizing Absorbent Pads
SDP
Catalog No. 22445780 Shop All Mopec Products
Change view
Click to view available options
Packaging Quantity:
100 Pads/Pk., 25 Pk./Cs.
Catalog No. Packaging Quantity
22-445-780 100 Pads/Pk., 25 Pk./Cs.
1 options
Catalog No. 22-445-780 Supplier Mopec™ Supplier No. BE092
Add to Cart
Add to Cart
The Formaway Formalin Neutralizing Absorbent Pad (F-NAP) was designed to reduce formaldehyde vapors while dissecting tissue, transferring tissue from containers of formalin, and transporting or storing tissue in containers of formalin, by absorbing and neutralizing formalin.
  • 3 x 4 in. pads
Specifications
Material Pre-treated Formalin Neutralizer
Type Formalin Absorbent
Packaging Quantity 100 Pads/Pk., 25 Pk./Cs.