Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lux Blox™ Mini Freestyle Set
SCI_ED

Catalog No. S81704
Change view
Click to view available options
Grade:
Pre-K to 12
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S81704 Pre-K to 12
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S81704 Supplier Lux Blox™ Supplier No. LUX66
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The perfect way for children to try out the world of Lux Blox

  • One-of-a-kind hinged pieces snap together to allow children o build anything their imaginations can dream up
  • Durable building block pieces connect on all 4 sides to allow for more flexible builds, creating toys where any part can move
  • Set includes instructions for multiple builds including a Lux Cycle, Robot, Monoplane, and Martian Lander
  • Supervised use is recommended for young children

Specifications

Color Neon Orange, Teal Blue, Black, and White
Grade Pre-K to 12
Age 6
Includes Instructions to build a Lux Cycle, Robot, Monoplane, Martian Lander and more! 66 Lux Pieces in Neon Orange, Teal Blue, Black and White plus accessories
Material ABS Plastic
Product Type Mini Freestyle Set
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.