Learn More
Medchemexpress LLC HY-P1593A 1mg Medchemexpress, Mini Gastrin I, human (TFA) CAS: Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYP1593A1MG
Encompass_Preferred
Medchemexpress, HY-P1593A 1mg Mini Gastrin I, human (TFA) CAS: Mini Gastrin I, human (TFA) is a shorter version of human gastrin, consists of amino acids 5-17 of the parent peptide. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-3070