Learn More
Fine Science Tools MINI GOLDSTEIN 1/2 RETRACTOR
SDP
Supplier:  Fine Science Tools 1700202
Encompass
Product: Mini-Goldstein Retractor - 3x3 Sharp/2cm/0.5cm Maximum Spread/ Teeth: 3x3 Sharp/ Teeth Length: 4mm/ Teeth Width: 5.5mm/ Maximum Spread: 0.5cm/ Alloy: Stainless Steel/ Length: 2cm/ Unit: Each
Catalog No. NC9054612