Learn More
Bulbman Inc MINI HALOGEN BULB T3 20W 12V
SDP
Catalog No. NC0477895
Encompass_Preferred
Osram Sylvania tungsten halogen quartz 20 watt, 12 volt standard low pressure clear finish UV-filter with transverse filament G4 bi-pin base 2000 hour lamp life
Catalog No. NC0477895 Supplier Bulbman Inc Supplier No. 107466
Osram Sylvania tungsten halogen quartz 20 watt, 12 volt standard low pressure clear finish UV-filter with transverse filament G4 bi-pin base 2000 hour lamp life