Learn More
IBI SCIENTIFIC 3P MINI I-BLUE PLSMID KT 100 PREP
Catalog No. NC0487042
Encompass_Preferred
DNA Purification Kit, I-Blue Mini Plasmid Kit (100-Prep), IBI Scientific, Sample Size: 1-7ml bacterial culture, Operation Time: 20min, Expectant Yield: up to 50ug of pure plasmid DNA, Elution Volume: 30-100ul, Format: Plasmid Spin Column.
Catalog No. NC0487042 Supplier IBI SCIENTIFIC 3P Supplier No. IB47171
DNA Purification Kit, I-Blue Mini Plasmid Kit (100-Prep), IBI Scientific, Sample Size: 1-7ml bacterial culture, Operation Time: 20min, Expectant Yield: up to 50ug of pure plasmid DNA, Elution Volume: 30-100ul, Format: Plasmid Spin Column.