Learn More
BURKLE INC Mini immersion cylinder, V2A, 1.69 oz. (50 ml), hxØ 7.09x1.26 in. (180x32 mm)
Catalog No. 5000025699
Encompass_Preferred
Mini immersion cylinder made of durable V2A stainless steel, with a capacity of 50 ml (1.69 oz) and compact dimensions (180 x 1.26 in. (32 mm)). Designed for sampling liquids in narrow or hard-to-reach places. Perfect for use in curved dipping tubes and barrels with narrow openings, ensuring precise and easy liquid sampling in laboratories, industry, and water treatment applications. Lowering rope sold separately.
Catalog No. 50-000-25699 Supplier BURKLE INC Supplier No. 53655000
Mini immersion cylinder made of durable V2A stainless steel, with a capacity of 50 ml (1.69 oz) and compact dimensions (180 x 1.26 in. (32 mm)). Designed for sampling liquids in narrow or hard-to-reach places. Perfect for use in curved dipping tubes and barrels with narrow openings, ensuring precise and easy liquid sampling in laboratories, industry, and water treatment applications. Lowering rope sold separately.