Learn More
Bio Rad Laboratories MINI INCUBATION TRAYS
Catalog No. 509010084
Encompass
Tray, Incubation; Bio-Rad; Trays accept 8-channel multichannel pipettors for simplified screening of microplate samples; Each tray has eight 10.5cm x 5mm channels; 100
Catalog No. 50-901-0084 Supplier Bio Rad Laboratories Supplier No. 1703903
Tray, Incubation; Bio-Rad; Trays accept 8-channel multichannel pipettors for simplified screening of microplate samples; Each tray has eight 10.5cm x 5mm channels; 100