Learn More
Research Products International Corp Mini-Incubator Shaker
SDP
Catalog No. 501976826
Encompass
The Incu-Shaker Mini is the most compact shaking incubator in its class. Despite the remarkably small footprint, a large 11.5 x 9.5 work space accepts several platforms and a variety of flasks, tubes and other common vessels.
The standard platform (included) features a non-slip, rubber coated surface, ideal for tissue culture flasks, Petri dishes and staining trays.
Catalog No. 50-197-6826 Supplier Research Products International Corp Supplier No. 248572
The Incu-Shaker Mini is the most compact shaking incubator in its class. Despite the remarkably small footprint, a large 11.5 x 9.5 work space accepts several platforms and a variety of flasks, tubes and other common vessels.
The standard platform (included) features a non-slip, rubber coated surface, ideal for tissue culture flasks, Petri dishes and staining trays.