Learn More
Alkali Scientific Incu-Shaker™ Mini Incubator Shaker, Touchscreen Control with Non-slip Rubber Mat, 115V
SDP
Catalog No. 502336199
Encompass_Preferred
The Incu-Shaker™ Mini features a compact design allowing for a smaller overall footprint and more work space on your lab bench. It has a temperature range of +5 to 60˚C and a speed range of 30 to 300 rpm. The speed can be adjusted in 1 rpm incruments. The platform dimensions of the shaking incubator is 9.5 x 11.5 in. or 23.5 x 29 cm with a lid height of 10.8 inches. It comes with a standard US electrical plug. A truly dependable machine that offers full functionality when incubating petri dishes, flasks, test tubes and other samples assays, the shaking incubator from Alkali Scientific is the right choice for your research.
Catalog No. 50-233-6199 Supplier Alkali Scientific Supplier No. RS7095
The Incu-Shaker™ Mini features a compact design allowing for a smaller overall footprint and more work space on your lab bench. It has a temperature range of +5 to 60˚C and a speed range of 30 to 300 rpm. The speed can be adjusted in 1 rpm incruments. The platform dimensions of the shaking incubator is 9.5 x 11.5 in. or 23.5 x 29 cm with a lid height of 10.8 inches. It comes with a standard US electrical plug. A truly dependable machine that offers full functionality when incubating petri dishes, flasks, test tubes and other samples assays, the shaking incubator from Alkali Scientific is the right choice for your research.