Learn More
METRIS INSTRUMENTS EAST LLC MINI-IR THERMOMETER W/CLIP 1:1
Catalog No. 50320896
Encompass_Preferred
Thermometer, IR; Metris Instruments; Mini; Temp. range: -33deg. to 230deg.C; +/-2%/+/-2deg.C accuracy; Fixed emissivity: 0.95; D:S: 1:1; Auto-off; Metal alloy case; LCD display; 3.5L x 0.5 in. D; 2 x LR44 batteries
Catalog No. 50-320-896 Supplier METRIS INSTRUMENTS EAST LLC Supplier No. TN002PC
Thermometer, IR; Metris Instruments; Mini; Temp. range: -33deg. to 230deg.C; +/-2%/+/-2deg.C accuracy; Fixed emissivity: 0.95; D:S: 1:1; Auto-off; Metal alloy case; LCD display; 3.5L x 0.5 in. D; 2 x LR44 batteries