Learn More
World Precision Instrument Mini Iris Scissors, 8cm, Curved, Sharp Tips
SDP
Supplier:  World Precision Instrument 503671
Encompass
Iris scissors were originally designed for ophthalmic procedures but are now used in a wide range of applications. The tips are sharp, and they come in curved, straight or angled styles.
Catalog No. 50-253-9464