Learn More
Thomas Scientific Mini Labroller Rotator, 120V
SDP
Catalog No. 501460047
Encompass
Designed for mixing samples in both a horizontal and vertical plane Compact design Wide selection of motions Noiseless operation Cold room compatible Dual format rotisseries Three easily interchangeable rotisseries Holds various size tubes: 36 x 1.5 / 2.0ml; 10 x 15ml/12 x 5ml; and 6 x 50ml The unit accepts a broad assortment of tube sizes as well as microplates. Safe for cold room or incubator use, this mixer is ideal for a wide range of applications in areas such as biochemistry, molecular biology, clinical testing and histochemistry. Compact and sleek design. A small footprint and efficient housing design conserve valuable bench space and make the Mini LabRoller Rotator suitable for any size laboratory. It has an ambient operating range of 4 to 65°C and will easily fit into most incubators. Lightweight and portable, the unit is easily moved from the bench to a biological or chemical hood. Simple construction allows for easy cleaning and decontamination. The Mini LabRoller Rotato
Catalog No. 50-146-0047 Supplier Thomas Scientific Supplier No. 1228P26
Designed for mixing samples in both a horizontal and vertical plane Compact design Wide selection of motions Noiseless operation Cold room compatible Dual format rotisseries Three easily interchangeable rotisseries Holds various size tubes: 36 x 1.5 / 2.0ml; 10 x 15ml/12 x 5ml; and 6 x 50ml The unit accepts a broad assortment of tube sizes as well as microplates. Safe for cold room or incubator use, this mixer is ideal for a wide range of applications in areas such as biochemistry, molecular biology, clinical testing and histochemistry. Compact and sleek design. A small footprint and efficient housing design conserve valuable bench space and make the Mini LabRoller Rotator suitable for any size laboratory. It has an ambient operating range of 4 to 65°C and will easily fit into most incubators. Lightweight and portable, the unit is easily moved from the bench to a biological or chemical hood. Simple construction allows for easy cleaning and decontamination. The Mini LabRoller Rotato