Learn More
Andwin Scientific MINI LABROLLER ROTATOR LABNET
SDP
Catalog No. NC9705412
Encompass_Preferred
Mini LabRoller Rotator with rotisserie for 36x1.5/2.0ml tubes, 1/EA. Simple, yet versatile, the Mini LabRoller Rotator is designed for mixing samples in both a horizontal and vertical plane. The unit accepts a broad assortment of tube sizes as well as microplates. Safe for cold room or incubator use, this mixer meets a variety of needs in areas such as biochemistry, molecular biology, clinical testing and histochemistry.
Catalog No. NC9705412 Supplier Andwin Scientific Supplier No. H5500
Mini LabRoller Rotator with rotisserie for 36x1.5/2.0ml tubes, 1/EA. Simple, yet versatile, the Mini LabRoller Rotator is designed for mixing samples in both a horizontal and vertical plane. The unit accepts a broad assortment of tube sizes as well as microplates. Safe for cold room or incubator use, this mixer meets a variety of needs in areas such as biochemistry, molecular biology, clinical testing and histochemistry.