Learn More
Sigma Aldrich Fine Chemicals Biosciences Mini LabRollerTM rotator A1EA
Catalog No. 502905698
Encompass
Simple yet versatile the Mini LabRoller Rotator is designed for mixing samples in both a horizontal and vertical plane. The Mini LabRoller Rotator is supplied with a rotisserie for 36 x 1.5 or 2.0 mL tubes. Two additional rotisseries are available separately.Compact design conserves valuable benchSimple construction allows for easy cleaning and decontamination Wide selection of motions from gentle rolling tumbling to end over end mixingCold room and incubator compatibleThree easily interchangeable rotisseries to hold various size tubes and platesOperating conditions 4 C - 65 C up to 85% RRH non-condensingWhen a rotisserie is inserted horizontally (tubes held vertically) an end over end motion is produced. The unit can then be used for simple rotating or rested on either end to create a tumbling motion. Placing the Mini LabRoller on the end with the power switch will create a more vigorous tumbling motion than resting it on the opposite end.A rocking
Catalog No. 50-290-5698 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z6745831EA
Simple yet versatile the Mini LabRoller Rotator is designed for mixing samples in both a horizontal and vertical plane. The Mini LabRoller Rotator is supplied with a rotisserie for 36 x 1.5 or 2.0 mL tubes. Two additional rotisseries are available separately.Compact design conserves valuable benchSimple construction allows for easy cleaning and decontamination Wide selection of motions from gentle rolling tumbling to end over end mixingCold room and incubator compatibleThree easily interchangeable rotisseries to hold various size tubes and platesOperating conditions 4 C - 65 C up to 85% RRH non-condensingWhen a rotisserie is inserted horizontally (tubes held vertically) an end over end motion is produced. The unit can then be used for simple rotating or rested on either end to create a tumbling motion. Placing the Mini LabRoller on the end with the power switch will create a more vigorous tumbling motion than resting it on the opposite end.A rocking