Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Supertek™ Mini Lamp Sockets, Set of 12
SCI_ED

Catalog No. S139446 Shop All Supertek Scientific Products
Change view
Click to view available options
Grade:
1+
Catalog No. Grade
S139446 1+
1 options

Catalog No. S139446

Supplier: Supertek™ PH90771/2S312

These Supertek™ Mini Lamp Sockets are used for low voltage experiments.

  • Set of 12
  • Use for low voltage experiments.

Specifications

Grade 1+
Format Set
Languages English
Subject Physical Science - Electricity
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.