Learn More
Research Products International Corp Portable Mini Light Box, Benchtop Light Source
SDP
Catalog No. 50999343
Encompass
Ideal light box for viewing micro-well plates, petri-dishes, growth and color reactions, gels, slides, and transparencies. Compact unit saves valuable bench space and stores easily in a drawer when not in use.
Mini-Light Box is battery operated and completely portable. Powered by six AAA batteries (included).
Catalog No. 50-999-343 Supplier Research Products International Corp Supplier No. 815500
Ideal light box for viewing micro-well plates, petri-dishes, growth and color reactions, gels, slides, and transparencies. Compact unit saves valuable bench space and stores easily in a drawer when not in use.
Mini-Light Box is battery operated and completely portable. Powered by six AAA batteries (included).