Learn More
Roboz Surgical Roboz Galilean Loupes, 3X Magnification, 340 mm, 1 ea
SDP
Catalog No. 5000575053
Encompass
These lightweight 3X Galilean surgical loupes are integrated into comfortable sports-style eyeglasses to provide clear, magnified visualization during precision procedures. Designed with a 340 mm working distance, they balance a 70 mm field of view with a 100 mm depth of field for optimal performance in clinical settings.
  • Weighs 60 grams for extended use comfort
  • Includes a double-hinge design for precise optical positioning
  • Features a flip-up mechanism for rapid switching between magnified and normal views
  • Wide field of view optics
Catalog No. 50-005-75053 Supplier Roboz Surgical Supplier No. ML655
These lightweight 3X Galilean surgical loupes are integrated into comfortable sports-style eyeglasses to provide clear, magnified visualization during precision procedures. Designed with a 340 mm working distance, they balance a 70 mm field of view with a 100 mm depth of field for optimal performance in clinical settings.
  • Weighs 60 grams for extended use comfort
  • Includes a double-hinge design for precise optical positioning
  • Features a flip-up mechanism for rapid switching between magnified and normal views
  • Wide field of view optics