Learn More
Electron Microscopy Sciences Mini Magnetic Stirrer, 1/EA
SDP
Catalog No. 502091019
Encompass_Preferred
Mini Magnetic Stirrer is an economical stirrer with a optimal magnetic coupling that ensures dependable stirring efficiency. The powerful magnet coupling to a high-quality stir bar and the unit ensures consistent accurate alignment which significantly reduces spin-outs during acceleration or deceleration of spin speeds and ensure maximum stirring performance. The mini stirrer is made from an ABS and PC blend that is chemical resistant and easy to clean. The low-profile design is lightweight for transporting and the small footprint saves space on a bench top. The mini stirrer offers quiet operation with low vibration. It is deal for routine stirring applications in general research, life science, industrial, quality analysis, and clinical lab settings. 2 High quality stir bars (35 x 9 mm) are included to ensure maximum stirring performance. Color: Gray/Blue.
Catalog No. 50-209-1019 Supplier Electron Microscopy Sciences Supplier No. 7236802
Mini Magnetic Stirrer is an economical stirrer with a optimal magnetic coupling that ensures dependable stirring efficiency. The powerful magnet coupling to a high-quality stir bar and the unit ensures consistent accurate alignment which significantly reduces spin-outs during acceleration or deceleration of spin speeds and ensure maximum stirring performance. The mini stirrer is made from an ABS and PC blend that is chemical resistant and easy to clean. The low-profile design is lightweight for transporting and the small footprint saves space on a bench top. The mini stirrer offers quiet operation with low vibration. It is deal for routine stirring applications in general research, life science, industrial, quality analysis, and clinical lab settings. 2 High quality stir bars (35 x 9 mm) are included to ensure maximum stirring performance. Color: Gray/Blue.