Learn More
MSC XPower Manufacturing Air Mover, 1/5 HP, 800 CFM, 4-Speed, 115V
Catalog No. 502835360
Encompass
This Mini Mighty Air Mover is a portable unit designed for drying tight spaces, such as inside crawl spaces and cabinets. It features a 1/5 HP motor, 800 CFM airflow, 4 speeds, and operates on 115 V. This air mover is equipped with a refillable scent cartridge, a 3-hour timer, and built-in power outlets for daisy-chain functionality.
  • Portable alternative to HVAC nebulizers.
  • Features a patented refillable scent-diffusing cartridge that quickly and efficiently fragrances the air.
  • Powerful and energy efficient.
  • Lightweight and stackable.
  • Includes a 3-hour timer.
  • Equipped with a 10 ft. power cord with wrap-around design.
  • Built-in power outlets for daisy chain with dual thermal protection.
  • Kickstand enables 4-angle drying positions.
  • cETLus compliant.
Catalog No. 50-283-5360 Supplier MSC Supplier No. 98942725
This Mini Mighty Air Mover is a portable unit designed for drying tight spaces, such as inside crawl spaces and cabinets. It features a 1/5 HP motor, 800 CFM airflow, 4 speeds, and operates on 115 V. This air mover is equipped with a refillable scent cartridge, a 3-hour timer, and built-in power outlets for daisy-chain functionality.
  • Portable alternative to HVAC nebulizers.
  • Features a patented refillable scent-diffusing cartridge that quickly and efficiently fragrances the air.
  • Powerful and energy efficient.
  • Lightweight and stackable.
  • Includes a 3-hour timer.
  • Equipped with a 10 ft. power cord with wrap-around design.
  • Built-in power outlets for daisy chain with dual thermal protection.
  • Kickstand enables 4-angle drying positions.
  • cETLus compliant.