Learn More
MSC XPOWER Scented Air Mover, 1/8 HP, 500 CFM, Ionizer
Catalog No. 502835245
Encompass
This XPOWER P-150N scented air mover features a 1/8 HP motor and 500 CFM airflow, designed to improve air quality and facilitate drying in tight spaces. It includes a negative ion generator and refillable scent cartridge to neutralize odors and eliminate airborne pollutants. With 3 speeds and 4 drying positions, this lightweight unit offers versatile performance.
  • Powerful 1/8 HP induction motor
  • Low 1.5 Amp draw; 500 CFM airflow
  • 3 speeds, 4-angle drying positions
  • 10 ft. power cord with wrap-around design
  • Negative ion generator, refillable scent cartridge
  • Stackable up to 4 units
  • Lightweight, durable polypropylene housing
  • ETL/CETL safety certified
  • Quiet operation
  • Eliminates airborne pollutants
Catalog No. 50-283-5245 Supplier MSC Supplier No. 98942857
This XPOWER P-150N scented air mover features a 1/8 HP motor and 500 CFM airflow, designed to improve air quality and facilitate drying in tight spaces. It includes a negative ion generator and refillable scent cartridge to neutralize odors and eliminate airborne pollutants. With 3 speeds and 4 drying positions, this lightweight unit offers versatile performance.
  • Powerful 1/8 HP induction motor
  • Low 1.5 Amp draw; 500 CFM airflow
  • 3 speeds, 4-angle drying positions
  • 10 ft. power cord with wrap-around design
  • Negative ion generator, refillable scent cartridge
  • Stackable up to 4 units
  • Lightweight, durable polypropylene housing
  • ETL/CETL safety certified
  • Quiet operation
  • Eliminates airborne pollutants