The XPOWER P-80A mini mighty air mover is designed for ventilating and drying, offering up to 600 CFM with an efficient 138-watt motor. Drawing only 1.2 Amps at its highest speed, it is manufactured with rugged injection mold technology, ideal for drying tight spaces such as crawl spaces, beneath kitchen counters, inside cabinets, and auto interiors.
- Powerful and energy efficient 138 watt induction motor
- Low 1.2A draw with 600 CFM maximum airflow capacity
- Three speeds with 4-angle drying positions (0°, 20° kickstand, 45° & 90°)
- Ten ft. power cord with wrap-around cord design
- Built-in power outlets for daisy chain with dual thermal protection
- Stackable up to 4 units high for easy storage
- Lightweight, compact and durable polypropylene (PP) plastic housing design
- ETL/CETL safety certified