Learn More
MSC XPOWER Air Mover, 600 CFM, 138 W, 3 Speeds, 4 Positions
Catalog No. 502835318
Encompass
This mighty air mover offers powerful, energy-efficient ventilation and drying with a 138-watt induction motor delivering up to 600 CFM airflow. It features three speeds and four-angle drying positions. Designed for convenience, it includes a 10 ft. power cord with wrap-around design, built-in power outlets for daisy-chaining, and is stackable. Its durable polypropylene housing makes it lightweight, compact, and ETL/CETL safety certified, making it ideal for drying tight spaces.
  • Energy-efficient 138 W induction motor
  • 600 CFM maximum airflow
  • Three speed settings
  • Four-angle drying positions
  • 10 ft. power cord with wrap-around design
  • Built-in power outlets for daisy-chaining
  • Stackable design for storage
  • Durable polypropylene housing
  • ETL/CETL safety certified
  • Suitable for drying tight spaces
Catalog No. 50-283-5318 Supplier MSC Supplier No. 98942667
This mighty air mover offers powerful, energy-efficient ventilation and drying with a 138-watt induction motor delivering up to 600 CFM airflow. It features three speeds and four-angle drying positions. Designed for convenience, it includes a 10 ft. power cord with wrap-around design, built-in power outlets for daisy-chaining, and is stackable. Its durable polypropylene housing makes it lightweight, compact, and ETL/CETL safety certified, making it ideal for drying tight spaces.
  • Energy-efficient 138 W induction motor
  • 600 CFM maximum airflow
  • Three speed settings
  • Four-angle drying positions
  • 10 ft. power cord with wrap-around design
  • Built-in power outlets for daisy-chaining
  • Stackable design for storage
  • Durable polypropylene housing
  • ETL/CETL safety certified
  • Suitable for drying tight spaces