Learn More
Electron Microscopy Sciences Mini Mixer
SDP
Catalog No. 50949177
Encompass_Preferred
Perfect for mixing in test tubes, micro centrifuge tubes and other containers which are too narrow for conventional mixers. The stirring rod will also flare out for use in larger spaces. It comes with a clear vinyl carrying case, 4 1/2"x 1/4"stirring rod, and two "AA" batteries.
Catalog No. 50-949-177 Supplier Electron Microscopy Sciences Supplier No. 7184910
Perfect for mixing in test tubes, micro centrifuge tubes and other containers which are too narrow for conventional mixers. The stirring rod will also flare out for use in larger spaces. It comes with a clear vinyl carrying case, 4 1/2"x 1/4"stirring rod, and two "AA" batteries.