Learn More
Research Products International Corp Mini Mixer 3D Shaker with Dimpled Mat for 1.5ml to 50ml Tubes, 10.5 x 7.5 Inches, 230 Volt Model for Foreign Countries
SDP
Catalog No. 501977179
Encompass
The MiniMixer combines the motions of an orbital and rocking shaker to produce a three dimensional and "nutating" gyration. Provides a gentle, thorough action without causing foaming. Ideal for re-suspending fragile DNA fragments and mixing of biological samples.
Even the smallest single platform configuration has 24% more platform work surface than traditional nutating mixers. These mixers feature maintenance free, brushless motors capable of loads up to 2 kilograms and are safe for use in cold rooms and incubators.
This mixer has an ambient operating range of +4°C up to 65°C allowing use in a cold room or incubator. Supplied with a dimpled rubber mat that holds tubes firmly in place. Accommodates variety of sample tube sizes from 1.5 ml up to 50ml.
Catalog No. 50-197-7179 Supplier Research Products International Corp Supplier No. 248567230
The MiniMixer combines the motions of an orbital and rocking shaker to produce a three dimensional and "nutating" gyration. Provides a gentle, thorough action without causing foaming. Ideal for re-suspending fragile DNA fragments and mixing of biological samples.
Even the smallest single platform configuration has 24% more platform work surface than traditional nutating mixers. These mixers feature maintenance free, brushless motors capable of loads up to 2 kilograms and are safe for use in cold rooms and incubators.
This mixer has an ambient operating range of +4°C up to 65°C allowing use in a cold room or incubator. Supplied with a dimpled rubber mat that holds tubes firmly in place. Accommodates variety of sample tube sizes from 1.5 ml up to 50ml.