Learn More
Chemglass Life Sciences Mini Nutating Rocker, 8" x 6" Platform, 120V
SDP
Catalog No. 501311305
Encompass
Mini Nutating Rocker, 8" x 6" Platform, 120V. Mini Nutating Rocker:The smallest in our line of rocker¿s, the CLS-2457-05 is specifically designed to fit inside the myTemp. The myTemp door is designed to seal around the CLS-2457-05¿s flat cord when the door is closed.3D (nutating) motionFixed speed mixing: 24rpmFixed ¿20¿ tilt8 x 6¿ platformIncludes two mats: one flat and one dimpled4lb load capacityCold-room and incubator safeCE-marked2 year warranty
Catalog No. 50-131-1305 Supplier Chemglass Life Sciences Supplier No. CLS245705
Mini Nutating Rocker, 8" x 6" Platform, 120V. Mini Nutating Rocker:The smallest in our line of rocker¿s, the CLS-2457-05 is specifically designed to fit inside the myTemp. The myTemp door is designed to seal around the CLS-2457-05¿s flat cord when the door is closed.3D (nutating) motionFixed speed mixing: 24rpmFixed ¿20¿ tilt8 x 6¿ platformIncludes two mats: one flat and one dimpled4lb load capacityCold-room and incubator safeCE-marked2 year warranty