Learn More
Bellco Glass, Inc. Mini-Oven,115V w/6 Position Rotisserie
SDP
Catalog No. 501946525
Encompass_Preferred
The Bellco Auto-blot Mini Hybridization Oven consists of an incubator with an integral rotisserie device. The rotisserie has the capacity for up to 4 large hybridization bottles or 8 small hybridization bottles. Alternatively, any combination of large or small bottles can be used. This Auto-blot Micro Hybridization Oven features an advanced temperature controller for precise control of oven temperature. 14”x17”x13” (35x42x32cm) Int: 7”x15”x8” (18x37x20cm) 115v. MADE IN THE USA
Catalog No. 50-194-6525 Supplier Bellco Glass, Inc. Supplier No. 793010110
The Bellco Auto-blot Mini Hybridization Oven consists of an incubator with an integral rotisserie device. The rotisserie has the capacity for up to 4 large hybridization bottles or 8 small hybridization bottles. Alternatively, any combination of large or small bottles can be used. This Auto-blot Micro Hybridization Oven features an advanced temperature controller for precise control of oven temperature. 14”x17”x13” (35x42x32cm) Int: 7”x15”x8” (18x37x20cm) 115v. MADE IN THE USA