Learn More
Apacor Mini Parasep™ SF
Catalog No. 22332023
Change view
Click to view available options
Quantity:
1000 Tests
Catalog No. Quantity
22-332-023 1000 Tests
1 options
This item is not returnable. View return policy
Catalog No. 22-332-023 Supplier Apacor Supplier No. 108000
Add to Cart
Edge
Add to Cart
This item is not returnable. View return policy
For the clean and efficient concentration of Helminth ova and larvae and Protozoa cysts and oocysts
  • Single use device for in vitro diagnostic use only
  • Totally enclosed/sealed process
  • Disposable device
  • Single use, eliminates cross contamination
  • Pre-filled systems available
  • Optimum sample recovery
  • Enhanced sample clarity
  • Rapid four step process
  • Human resources optimised
  • Reduces human error
Specifications
Color Clear, Green
Description Fecal Parasite Concentrators – Solvent Free
Quantity 1000 Tests
Volume (Metric) 15 mL