Learn More
Research Products International Corp Mini PCR Plate Centrifuge
SDP
Catalog No. 501977012
Encompass
The PCR Plate Centrifuge is the most compact and economical instrument available for spinning PCR plates. Significantly improves yields when used before and after thermal cycling to spin down condensation droplets. Accommodates two skirted or nonskirted plates designed for PCR.
PCR Plate Spinner is an excellent option for those looking for a personal-sized centrifuge. Quickly spin down droplets and condensation. Use before and after thermal cycling to increase PCR yield. Accepts skirted, non-skirted and all standard PCR plates. Less than 1/4 the size of most mini PCR plate centrifuges.
Mini Plate Spinner is the first centrifuge designed specifically for quick and easy centrifugation of samples in PCR plates or microplates. Centrifuges that accept microplates are typically larger units that take up valuable space on the bench. The unique design of the Mini Plate Spinner results in a compact centrifuge that has an extremely small footprint - just 6 x 6 inches.
Catalog No. 50-197-7012 Supplier Research Products International Corp Supplier No. 141370
The PCR Plate Centrifuge is the most compact and economical instrument available for spinning PCR plates. Significantly improves yields when used before and after thermal cycling to spin down condensation droplets. Accommodates two skirted or nonskirted plates designed for PCR.
PCR Plate Spinner is an excellent option for those looking for a personal-sized centrifuge. Quickly spin down droplets and condensation. Use before and after thermal cycling to increase PCR yield. Accepts skirted, non-skirted and all standard PCR plates. Less than 1/4 the size of most mini PCR plate centrifuges.
Mini Plate Spinner is the first centrifuge designed specifically for quick and easy centrifugation of samples in PCR plates or microplates. Centrifuges that accept microplates are typically larger units that take up valuable space on the bench. The unique design of the Mini Plate Spinner results in a compact centrifuge that has an extremely small footprint - just 6 x 6 inches.