Learn More
World Precision Instrument Mini Peristaltic Pump-504011
SDP
Catalog No. 502539428
Encompass
This compact, lightweight peristaltic pump fits just about anywhere. It can be mounted directly on the bench, in a regular rack or to a post. The speed can be adjusted from 1 to 50 rpm. With recommended silicone tubing, the volume can be set from 0.06 to 14.0 mL/min. The MiniStar™ also features a hand held remote control that allows you to start and stop the pump, purge or adjust its speed and direction.
Catalog No. 50-253-9428 Supplier World Precision Instrument Supplier No. 504011
This compact, lightweight peristaltic pump fits just about anywhere. It can be mounted directly on the bench, in a regular rack or to a post. The speed can be adjusted from 1 to 50 rpm. With recommended silicone tubing, the volume can be set from 0.06 to 14.0 mL/min. The MiniStar™ also features a hand held remote control that allows you to start and stop the pump, purge or adjust its speed and direction.