Learn More
Research Products International Corp Mini-Pipettor, 100ul, Blue
SDP
Catalog No. 501367750
Encompass
Ideal size Lil’Pet is compact and features optimum size to facilitate ease of handling. Engineered for precise air displacement pipetting with two stage plunger and allows last drop dispensing. Available in a full range of volumes (5µl to 1000µl) and color plunger options. Unique tip cone accepts both micro tips (200µl) and ultra micro tip (up to 25µl). Built from sturdy molded polypropylene, the Lil’Pet handles thousands of pipettings and is fully autoclavable. CE marked.
Catalog No. 50-136-7750 Supplier Research Products International Corp Supplier No. 347025
Ideal size Lil’Pet is compact and features optimum size to facilitate ease of handling. Engineered for precise air displacement pipetting with two stage plunger and allows last drop dispensing. Available in a full range of volumes (5µl to 1000µl) and color plunger options. Unique tip cone accepts both micro tips (200µl) and ultra micro tip (up to 25µl). Built from sturdy molded polypropylene, the Lil’Pet handles thousands of pipettings and is fully autoclavable. CE marked.