Learn More
Hoefer, Inc Hoefer™ SUB10 Mini-Plus Submarine Gel Electrophoresis Unit
SDP
Catalog No. NC2741102
Encompass_Preferred
Ideal for routine preparatory and analytical electrophoresis techniquesFeatures and BenefitsColor-coded and height adjustable combs—easily identify comb thickness at a glance and control well depthColored loading strips—for easy well detection when loadingCompact tank—reduces the buffer volume required to cover the gel, providing greater control over the voltage gradient and run-timeUV-transparent running tray—allows the user to image the gel without risk of damage due to handlingSide handles—for safe and easy transportation around the laboratoryIncludes:Buffer Chamber Assembly; Safety Lid; High Voltage Leads; Running Tray with Silicone Gasket; 1 mm thick, 16-well combs, 2 pcs; Colored Loading StripsSpecifications:Gel Dimensions (w x l): 10 x 11.5 cmMaximum Buffer Volume: 450 mlMaximum Power Settings: 300 V, 200 mA, 60 WMaximum Sample Capacity: 80Unit Dimensions (w x l x h): 16.5 x 23 x 6.5 cmSafety Certifications: EN61010-1, CE
Catalog No. NC2741102 Supplier Hoefer, Inc Supplier No. SUB10
Ideal for routine preparatory and analytical electrophoresis techniquesFeatures and BenefitsColor-coded and height adjustable combs—easily identify comb thickness at a glance and control well depthColored loading strips—for easy well detection when loadingCompact tank—reduces the buffer volume required to cover the gel, providing greater control over the voltage gradient and run-timeUV-transparent running tray—allows the user to image the gel without risk of damage due to handlingSide handles—for safe and easy transportation around the laboratoryIncludes:Buffer Chamber Assembly; Safety Lid; High Voltage Leads; Running Tray with Silicone Gasket; 1 mm thick, 16-well combs, 2 pcs; Colored Loading StripsSpecifications:Gel Dimensions (w x l): 10 x 11.5 cmMaximum Buffer Volume: 450 mlMaximum Power Settings: 300 V, 200 mA, 60 WMaximum Sample Capacity: 80Unit Dimensions (w x l x h): 16.5 x 23 x 6.5 cmSafety Certifications: EN61010-1, CE