Learn More
Research Products International Corp Mini-Pipettor, 50ul, YLW
SDP
Supplier:  Research Products International Corp 347024
Encompass
Compact and engineered for precision dispensing, the sturdy Minipet air-displacement microPPTtes come in a range of volumes (5ul to 200ul) and color options. Most important, the MiniPet microPPTte provides as low as +/-3% accuracy and 2% precision at full stroke. Built from sturdy molded PLAS, the air-displacement MiniPet microPPTte handles thousands of PPTtings. Length: 9.53cm Handle Diameter: 1.37cm Polypropylene (Disposable Tip Only)Shipping Weight: 1 lbsShipping Dimensions: 2.00 x 4.50 x 1.00Product usually ships within 24 hours.Research or further manufacturing use only, not for food or drug use.
Catalog No. 50-222-9468