Learn More
Bio Rad Laboratories MINI-PROTEAN 3 COMB
Catalog No. 509012302
Encompass
Comb; Bio-Rad; Mini-Protean; Prep/2-D well; Contains a reference well; 0.75mm
Catalog No. 50-901-2302 Supplier Bio Rad Laboratories Supplier No. 1653356
Comb; Bio-Rad; Mini-Protean; Prep/2-D well; Contains a reference well; 0.75mm