Learn More
Bio Rad Laboratories MINI-PROTEAN TETRA CELL
Catalog No. 509010535
Encompass
Cell, Tetra; Bio-Rad; Mini-Protean; 10-well; 0.75mm thick; 4-gel system w/combs, glass plates, casting stands and frames, sample loading guide, electrode assembly, companion running module, tank, lid w/power cables, mini cell buffer dam
Catalog No. 50-901-0535 Supplier Bio Rad Laboratories Supplier No. 1658000
Cell, Tetra; Bio-Rad; Mini-Protean; 10-well; 0.75mm thick; 4-gel system w/combs, glass plates, casting stands and frames, sample loading guide, electrode assembly, companion running module, tank, lid w/power cables, mini cell buffer dam