Learn More
Bio Rad Laboratories MINI-PROTEAN TETRA CELL
Supplier:  Bio Rad Laboratories 1658003
Encompass
Cell, Tetra; Bio-Rad; Mini-Protean; 10-well; 1.0mm thickness; 2-gel system includes 5 combs, 5 sets of glass plates, casting stand, 2 casting frames, sample loading guide, electrode assembly, tank, lid w/power cables, mini cell buffer dam
Catalog No. 50-901-0124