Learn More
Bio Rad Laboratories MINI-PROTEAN TETRA CELL CAST
Catalog No. 509010493
Encompass
Module, Casting; Bio-Rad; Mini-Protean; Tetra Cell; 10-well; 1.5mm; Includes 2 casting stands, 4 casting frames, 5 combs, 5 sets of plates, sample loading guide
Catalog No. 50-901-0493 Supplier Bio Rad Laboratories Supplier No. 1658021
Module, Casting; Bio-Rad; Mini-Protean; Tetra Cell; 10-well; 1.5mm; Includes 2 casting stands, 4 casting frames, 5 combs, 5 sets of plates, sample loading guide