Learn More
Bio Rad Laboratories MINI-PROTEAN TETRA CELL FOR RE
Catalog No. 509010497
Encompass
Cell, Tetra; Bio-Rad; Mini-Protean; For Ready Gel precast gels; 4-gel system includes electrode assembly, companion running module, tank, lid w/power cables, mini cell buffer dam
Catalog No. 50-901-0497 Supplier Bio Rad Laboratories Supplier No. 1658004
Cell, Tetra; Bio-Rad; Mini-Protean; For Ready Gel precast gels; 4-gel system includes electrode assembly, companion running module, tank, lid w/power cables, mini cell buffer dam