Learn More
Bio Rad Laboratories MINI-PROTEAN TETRA FOR PRECAST
Catalog No. 509010596
Encompass
Cell, Tetra; Bio-Rad; Mini-Protean; For Ready Gel precast gels and Mini Trans-Blot module; 4-gel system includes electrode assembly, companion running module, tank, lid w/power cables, mini cell buffer dam, Mini Trans-Blot module
Catalog No. 50-901-0596 Supplier Bio Rad Laboratories Supplier No. 1658030
Cell, Tetra; Bio-Rad; Mini-Protean; For Ready Gel precast gels and Mini Trans-Blot module; 4-gel system includes electrode assembly, companion running module, tank, lid w/power cables, mini cell buffer dam, Mini Trans-Blot module