Learn More
Bio Rad Laboratories MINI-PROTEAN TGX 4-15% 10W 10
Catalog No. 50950182
Encompass
Mini-PROTEAN TGX extended shelf-life gels are precast polyacrylamide gels for vertical SDS-PAGE and native PAGE protein separations. Gels 8.6 cm x 7.2 cm x 1.0 mm (W x L x thickness) are supplied in a 10.0 x 8.5 cm cassette.
Catalog No. 50-950-182 Supplier Bio Rad Laboratories Supplier No. 4561083
Mini-PROTEAN TGX extended shelf-life gels are precast polyacrylamide gels for vertical SDS-PAGE and native PAGE protein separations. Gels 8.6 cm x 7.2 cm x 1.0 mm (W x L x thickness) are supplied in a 10.0 x 8.5 cm cassette.