Learn More
Bio Rad Laboratories MINI-PROTEAN TGX STAIN FREE 4-
Catalog No. NC1085833
Encompass
Use these Mini-PROTEAN TGX Stain-Free Protein Gels for separation of polypeptides 2–400 kD. Long-life TGX, Tris-Glycine eXtended, Stain-Free Gels have a novel formulation, which allows rapid fluorescent detection of proteins with Bio-Rad’s stain-free enabled imaging systems. TGX Stain-Free Gels can be used for both standard denaturing protein separations as well as native electrophoresis. Run up to 4 Mini-PROTEAN TGX Stain-Free Protein Gels in 35 min using the Mini-PROTEAN Tetra Cell.
Catalog No. NC1085833 Supplier Bio Rad Laboratories Supplier No. 4568096
Use these Mini-PROTEAN TGX Stain-Free Protein Gels for separation of polypeptides 2–400 kD. Long-life TGX, Tris-Glycine eXtended, Stain-Free Gels have a novel formulation, which allows rapid fluorescent detection of proteins with Bio-Rad’s stain-free enabled imaging systems. TGX Stain-Free Gels can be used for both standard denaturing protein separations as well as native electrophoresis. Run up to 4 Mini-PROTEAN TGX Stain-Free Protein Gels in 35 min using the Mini-PROTEAN Tetra Cell.