Learn More
Supply Solutions cOmplete™, Mini Protease Inhibitor Cocktail, vial of 25 tablets
Supplier:  Supply Solutions SIGMA 11836153001
Encompass
Tablets provided in a glass vial. cOmplete, Mini Protease Inhibitor Tablets, inhibit a broad spectrum of serine, cysteine, and metalloproteases, as well as calpains.
Catalog No. 50-161-3322