Learn More
Thermo Scientific Drain Tubing, Mini Pump, Silicone
Supplier:  Thermo Scientific 842312052411
Encompass
Silicone peristaltic pump tubing for laboratory fluid transfer applications; flexible tubing interfaces with pump rotor for precise, contamination-free liquid handling including analytical testing and sample preparation.
Catalog No. NC1628605