Learn More
Research Products International Corp Mini Rad-Can Benchtop Container, 3/8"" Thick
SDP
Catalog No. 501977835
Encompass
The compact benchtop container shield is designed to fit easily on a laboratory bench or inside a fume hood. Constructed from durable acrylic for effective beta radiation containment, it features a hinged lid for easy access and is ideal for disposing of contaminated tips and tubes. Safe for freezer storage down to -70°C, this container provides a secure and reliable solution for handling and storing radioactive materials.
Catalog No. 50-197-7835 Supplier Research Products International Corp Supplier No. BRC03
The compact benchtop container shield is designed to fit easily on a laboratory bench or inside a fume hood. Constructed from durable acrylic for effective beta radiation containment, it features a hinged lid for easy access and is ideal for disposing of contaminated tips and tubes. Safe for freezer storage down to -70°C, this container provides a secure and reliable solution for handling and storing radioactive materials.