Learn More
AVANTOR MASTERFLEX (VWR LLC) Mini Tubing Clamps for up to 1/4"" Tubing, On/Off, Acetal; 25/Pk
Catalog No. NC0192945
Encompass
Mini Tubing Clamps for up to 1/4" Tubing, On/Off, Acetal; 25/PkDurable Celcon plastic clamp is autoclavable; won't rust, corrode, or distort. Accepts flexible tubing. Mini clamp accept up to 1/4" OD tubing and provides on/off control. The standard clamp accepts up to 1/2" OD tubing and features twelve-position ratchet control to adjust flow from fully-open to complete-closure.
Catalog No. NC0192945 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX0683301
Mini Tubing Clamps for up to 1/4" Tubing, On/Off, Acetal; 25/PkDurable Celcon plastic clamp is autoclavable; won't rust, corrode, or distort. Accepts flexible tubing. Mini clamp accept up to 1/4" OD tubing and provides on/off control. The standard clamp accepts up to 1/2" OD tubing and features twelve-position ratchet control to adjust flow from fully-open to complete-closure.