Learn More
Cole-Parmer Digi-Sense Panel-Mount Miniconnector, Type-T, Round, Female, 1/Ea
Catalog No. 501961550 Shop All Cole-Parmer Products
Encompass_Preferred
Mount connectors to your panel using the two screws located on the front of the jack. Each connector is ANSI color-coded by thermocouple type: black (J), yellow (K), or blue (T). Choose between flat-bladed miniconnectors and round-pinned standard connectors.
Catalog No. 50-196-1550 Supplier Cole-Parmer Supplier No. 1852723
Mount connectors to your panel using the two screws located on the front of the jack. Each connector is ANSI color-coded by thermocouple type: black (J), yellow (K), or blue (T). Choose between flat-bladed miniconnectors and round-pinned standard connectors.