Learn More
Bellco Glass, Inc. Mini Roll-In Inc,230V
SDP
Catalog No. 501946060
Encompass_Preferred
Bellco Mini Roll-In Incubators have 21 cubic feet of volume and are designed for use with Bellco's Cell Production Roller Apparatus (CPRA). The flush-to-floor design allows easy movement of the CPRA in and out of the chamber. Two internal convenience outlets.. MADE IN THE USA
Catalog No. 50-194-6060 Supplier Bellco Glass, Inc. Supplier No. 772865220
Add to Cart
Add to Cart
Request a Quote
Bellco Mini Roll-In Incubators have 21 cubic feet of volume and are designed for use with Bellco's Cell Production Roller Apparatus (CPRA). The flush-to-floor design allows easy movement of the CPRA in and out of the chamber. Two internal convenience outlets.. MADE IN THE USA