Learn More
Electron Microscopy Sciences Mini Rotator, 120 V
SDP
Catalog No. 5027304
Encompass_Preferred
The mini-rotator is a small, compact, rotator for micro centrifuge tubes and micro test tubes. It is heavy duty and has a permanent-magnet motor which maintains constant speed between 2-80 rpm. Mixing action is adjustable from a gentle slosh to a rapid agitation. It is ideal for use on a lab bench, in a small incubator or refrigerator, in a biological cabinet, or in a fume hood.
. For optimum mixing, the chemical resistant cabinet is fixed at a 20 degree angle. Unit comes complete with an aluminum disk head with spring clips to hold 15 each for both 5-10 mm and 12-14 mm tubes.
Catalog No. 50-273-04 Supplier Electron Microscopy Sciences Supplier No. 6510010
The mini-rotator is a small, compact, rotator for micro centrifuge tubes and micro test tubes. It is heavy duty and has a permanent-magnet motor which maintains constant speed between 2-80 rpm. Mixing action is adjustable from a gentle slosh to a rapid agitation. It is ideal for use on a lab bench, in a small incubator or refrigerator, in a biological cabinet, or in a fume hood.
. For optimum mixing, the chemical resistant cabinet is fixed at a 20 degree angle. Unit comes complete with an aluminum disk head with spring clips to hold 15 each for both 5-10 mm and 12-14 mm tubes.